Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6591

Sigma-Aldrich

JMJD2 (888-1023) GST tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

JHDM3A, Jumonji Domain Containing 2A, KDM4A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

42.2 kDa (888-1023 aa, NT GST Tag)

packaging

pkg of 20 and 50 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM4A(9682)

General description

Tudor domains are small protein structural motifs of about ~50 amino acids related to the “royal family” of methyl readers, which also includes chromo, MBT, PWWP, and Agenet-like domains. Tudor domains occur either alone, in tandem, or with other domains and are found in many proteins that are involved in RNA metabolism, germ cell development, transposon silencing, DNA damage response, histone modification, and chromatin remodeling. The tudor domains recognize symmetric methylated arginine or methylated lysine residues. JMJD2A catalyzes the demethylation of trimethylated histone H3 at lysine residues 9 and 36 (H3K9me3 and H3K36me3). However, the tudor domain of this protein has been shown to bind histone H3K4me, H3K9me3, and H3K20me2/3. Like other JmjC protein hydroxylase family members, JMJD2A is an α-ketoglutarate-dependent Fe (II) oxygenase. This product contains the tandem tudor domains of JMJD2A.

Physical form

50 mM Tris-HCl, pH 8.0, 150 mM sodium chloride, and 20% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service