Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6589

Sigma-Aldrich

SMN (1472-1613) GST tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Component of Gems 1, Gemin-1, Survival Motor Neuron Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

42.6 kDa (1472-1613 aa, NT GST Tag)

packaging

pkg of 20 μg
pkg of 50 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Tudor domains are small protein structural motifs of ~50 amino acids related to the “Royal family” of methyl readers, which also includes chromo, MBT, PWWP, and Agenet-like domains. Tudor domains occur either alone, in tandem, or with other domains and are found in many proteins that are involved in RNA metabolism, germ cell development, transposon silencing, DNA damage response, histone modification and chromatin remodeling. The tudor domains recognize symmetric methylated arginine or methylated lysine residues. The Survival of Motor Neurons (SMN) protein participates in RNA splicing. The Tudor domain of SMN recognizes and binds methylated Sm proteins, which bind small nuclear RNA. SMN is encoded in humans by two separate genes, SMN1 and SMN2, which differ by one base in exon 7. In motor neuron cells, approximately 90% of the SMN2 transcripts are spliced to exclude exon 7. The SMN2 transcripts without exon 7 are less stable than SMN1 transcripts. Consequently, defects in human SMN1 result in the death of motor neuron cells and spinal muscular atrophy, which is the leading genetic cause of infantile death. This protein product contains the tudor domain region of SMN. The sequence of this region is identical in both the SMN1 and the SMN2 genes.

Physical form

50 mM Tris-HCl, 150 mM sodium chloride, pH 8.0, containing 20% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service