Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6585

Sigma-Aldrich

Histone H2B (1-123) from Xanopus sp.

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

HIST1H2BA, Histone H2B type 1-A, TSH2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

Xenopus laevis

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

lyophilized powder

mol wt

13.7 kDa (1-123 aa)

packaging

pkg of 100 and 250 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... H2B1A(255626)

General description

H2A is a core component of nucleosome. A nucleosome is the basic repeating unit of chromatin in which 146 base pairs of DNA wrap twice around an octamer of histones. The octamer is composed of two of each histone H2A, H2B, H3, and H4. DNA accessibility is regulated via a complex set of post-translational modifications of these histones, also called histone code, and nucleosome remodeling. Histones H2A and H2B form a dimer. Histones H3 and H4 form a tetramer. The combination of two H2A/H2B dimers and one H3/H4 tetramer create the nucleosome core. Histone H2B undergoes many modifications which include acetylation, phosphorylation, ubiquitylation, sumoylation, and biotinylation that are important for regulation of gene transcription. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability.

Physical form

Lyophilized powder. Recommended buffer is 50 mM sodium phosphate, pH 7.2, containing 300 mM sodium chloride, 1 mM DTT, 1 mM EDTA and 20% glycerol.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service