Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6571

Sigma-Aldrich

BAZ2B (2064-2168) GST tag human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

Bromodomain Adjacent to Zinc Finger Domain 2B, KIAA1476, WALp4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

liquid

mol wt

40.2 kDa (2064-2168 aa + NT GST Tag)

packaging

pkg of 100 μg
pkg of 20 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BAZ2B(29994)

General description

The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. BAZ2B is expressed in heart, skeletal, testis, and pancreatic tissues. A rare allele of BAZ2B has been identified to be a predictor of Sudden Cardiac Death.4 The full-length protein contains several DNA-targeting domains, including a methyl-CpG binding domain, a DNA-binding DDT domain, and a tandem PHD-bromodomain. This protein product contains the bromodomain of BAZ2B.

Physical form

50 mM Tris, pH 7.5, containing 500 mM sodium chloride, 5% glycerol, and 5 mM β-mercaptoethanol.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service