Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6563

Sigma-Aldrich

BrdT (257-382) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

CT9, Cancer/testis antigen 9, RING3-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

liquid

mol wt

14.9 kDa (126 aa, 257-382 aa + NT His Tag)

packaging

pkg of 1 mg
pkg of 100 μg
pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRDT(676)

General description

BrdT is similar to the RING3 protein family. It possesses 2 bromodomain motifs and a PEST sequence. The bromodomain is found in proteins that regulate transcription. BrdT drives a meiotic and post-meiotic gene expression program. It also controls the genome-wide post-meiotic genome reorganization that occurs after histone hyperacetylation in elongating spermatids. It may play a role in the transcriptional regulation of spermatogenesis. It also seems to have a structural ATP-independent role in the reorganization of acetylated chromatin. Therecombinant protein includes Bromo 1 domain (257-382 aa) with N-terminal His-tag.

Physical form

rh-BrdT is supplied as a solution in 50 mM Tris, pH 8.0, 150 mM NaCl and 10% Glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service