Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6545

Sigma-Aldrich

Meteorin-like from mouse

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Cometin, Metrnl, Subfatin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

~350 kDa (Mouse meteorin-like 46-311 is fused at the N-terminus to a FLAG0174-tag.)

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Metrnl(210029)

General description

Meteorin-like (cometin; subfatin) is a potent new neurotrophic factor promoting neurite outgrowth from dorsal root ganglion cells. Meteorin-like is also a novel adipokine expressed by adipose tissue being downregulated upon caloric restriction. Meteorin-like is secreted by muscles during exercise and converts white adipose cells into brown fat tissue.

Physical form

Lyophilized in PBS.

Reconstitution

Reconstitute with 100 μsterile distilled water.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service