Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6535

Sigma-Aldrich

RBP4 from rat

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)

Synonym(s):

PRBP, Plasma retinol-binding protein, RBP, Retinol Binding Protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

rat

recombinant

expressed in HEK 293 cells

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

monomer ~230 kDa (Rat RBP4 aa 19-201 is fused at the N-terminus to a FLAG0174-tag.)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<0.1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... RBP4(25703)

General description

Retinol binding protein 4 (RBP4; RBP) is a 21 kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol (vitamin A) to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 (GLUT4) suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity, and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function.

Physical form

0.2 μm-filtered solution in PBS, pH 7.2.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service