Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6531

Sigma-Aldrich

NMNAT3 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

EC 2.7.7.1, EC 2.7.7.18, NMN Adenylyltransferase 3, NaMN Adenylyltransferase 3, Nicotinamide Mononucleotide Adenylyl-transferase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

monomer ~270 kDa (Human full-length NMNAT3 aa 1-252 is fused at the N-terminus to a His-tag.)

packaging

pkg of 10 and 50 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NMNAT3(349565)

General description

This is the Mitochondrial NMNAT isoform. It catalyzes the formation of NAD+ from nicotinamide mononucleotide (NMN) and ATP. It can also use the deamidated form of nicotinic acid mononucleotide (NAMN) as substrate with the same efficiency. It can use tiazofurin monophosphate as substrate. It can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD+. For the pyrophosphorolytic activity, mitochondrial NMNAT isoform can use NAD (+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. It fails to cleave phosphorylated dinucleotides NADP+, NADPH and NAADP+. It protects against axonal degeneration following injury.

Physical form

1 mg/mL in 50 mM sodium phosphate, pH 8.0 containing 300 mM sodium chloride, 1 mM DTT and 10% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service