Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6523

Sigma-Aldrich

ApoJ/Clusterin from mouse

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

AAG4, Apolipoprotein J, CLI, CLU, KUB1, MGC24903, SGP-2, SGP2, TRPM-2, TRPM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

~45 kDa (Mouse nuclear Clusterin aa 22- 448 is fused at the C-terminus to a His-tag.)

packaging

pkg of 10 μg
pkg of 50 μg

concentration

1 mg/mL

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CLU(12759)

General description

Native Apolipoprotein J (ApoJ), also named Clusterin, is a heavily glycosylated, 75-80 kDa disulfide-linked heterodimeric protein. Despite being cloned since 1989, no genuine function has been attributed to ApoJ so far. The protein has been reportedly implicated in several diverse physiological processes such as sperm maturation, lipid transportation, complement inhibition, tissue remodeling, membrane recycling, cell-cell and cell-substratum interactions, stabilization of stressed proteins in a folding-competent state and promotion or inhibition of apoptosis. ApoJ gene is differentially regulated by cytokines, growth factors and stress-inducing agents. Clusterin is up- or down regulated on the mRNA or protein level in many pathological and clinically relevant situations including cancer, organ regeneration, infection, Alzheimer disease, retinitis pigmentosa, myocardial infarction, renal tubular damage, autoimmunity and others.

Physical form

1 mg/mL of 0.2 μm-filtered solution in 55 mM Tris-Cl, pH 8.2, containing 150 mM NaCl.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service