Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6515

Sigma-Aldrich

Hemopexin from rat serum

≥95% (SDS-PAGE)

Synonym(s):

Beta-1B-glycoprotein, HPX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat serum

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

57 kDa

packaging

pkg of 100 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... HPX(58917)

General description

Hemopexin is a single-chain protein belonging to the family of blood transport proteins. It binds to heme released into the bloodstream during the degradation process. Recent studies have demonstrated that hemopexin acts as an extracellular antioxidant against hemoglobin-mediated damage in inflammation. Hemopexin protects against heme toxicity and conserves and recycles iron. Abnormal levels of hemopexin are associated with hemolytic anemia, chronic neuromuscular disease, and acute intermittent porphyria.

Physical form

Lyophilized from 11.9 mM phosphate, 137 mM NaCl, 2.7 mM KCl, pH 7.4.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service