Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6502

Sigma-Aldrich

LEPR/CD295 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

DKFZp686B1731, LEP-R, Leptin receptor, OB-R, OBR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 121 kDa (This protein is fused with Fc fragment of human IgG1 at the C-terminus.)
observed mol wt 150-165 kDa by SDS-PAGE (reducing) (Phe22 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LEPR(3953)

General description

Leptin receptor (LEPR), also known as LEP-R, cluster of differentiation 295 (CD295), OB-R and B219, is a single-transmembrane-domain receptor of the gp130 family of cytokine receptors. Leptin receptor exists as homodimer and binds Leptin with high affinity, thus mediates the biological function of the adipocyte-specific hormone Leptin. LEPR is a receptor for leptin (an adipocyte-specific hormone that regulates body weight), and is involved in the regulation of fat metabolism, as well as in a novel hematopoietic pathway that is required for normal lymphopoiesis. Mutations in this protein have been associated with obesity and pituitary dysfunction. Interaction of leptin and leptin receptor is crucial for body weight and bone mass regulation in mammals through hypothalamic effects on satiety and energy expenditure. Meanwhile, research data supports a leptin receptor activation model based on ligand-induced conformational changes.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service