Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6497

Sigma-Aldrich

IL7Ra/CD127 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 51 kDa (This protein is fused with Fc fragment of human IgG1 at the C-terminus.)
observed mol wt 66-90 kDa (DTT-reduced. Protein migrates due to glycosylation. Glu 21 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL17Ra(23765)

General description

Interleukin-7 receptor subunit α (IL7Ra) also known as CD antigen CD127, which belongs to the type I cytokine receptor family and type 4 subfamily. IL7Ra /CD127 contains 1 fibronectin type-III domain. IL7Ra /CD127 is expressed on various cell types, including naive and memory T cells and many others. IL7Ra /CD127 is receptor for interleukin-7 and also acts as a receptor for thymic stromal lymphopoietin (TSLP). The IL7 receptor is a heterodimer of IL7R and IL2RG. The TSLP receptor is a heterodimer of CRLF2 and IL7R.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service