Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6486

Sigma-Aldrich

TFPI2 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

PP5, REF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (N-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 22.7 kDa
observed mol wt 26-32 kDa (DTT-reduced. Asp 23 is the predicted N-terminal .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TFPI2(7980)

General description

Tissue Factor Pathway Inhibitor 2 (TFPI­2), also known as placental protein 5 (PP5) and retinal pigment epithelial cell factor 1 (REF­1), is a member of the Kunitz-type serine proteinase inhibitor family, is a structural homologue of tissue factor pathway inhibitor (TFPI). TFPI2 is highly abundant in the full-term placenta and widely expressed in umbilical vein endothelial cells, liver, placenta, heart, pancreas, and maternal serum at advanced pregnancy. TFPI­2 may play a role in the regulation of plasmin-mediated matrix remodeling. Inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa and has no effect on thrombin. Reduced synthesis of TFPI-2 has been related to numerous pathophysiological processes such as inflammation, angiogenesis, atherosclerosis, retinal degeneration and tumor growth / metastasis.

Physical form

Lyophilized from 0.22 μm filtered solution in 20 mM MES and 100 mM NaCl, pH 6.5. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangli Wang et al.
Scientific reports, 8(1), 14402-14402 (2018-09-28)
TFPI-2 has been recognized as a potent tumor suppressor gene. Low expression of TFPI-2 results in enhanced growth and metastasis of a variety of human tumors. In the present study, we investigated the mechanism responsible for the tumor suppressive effect
Muraly Puttabyatappa et al.
Endocrinology, 158(1), 109-120 (2016-11-05)
Increased proteolytic activity is a key event that aids in breakdown of the follicular wall to permit oocyte release. How the protease activity is regulated is still unknown. We hypothesize that tissue factor pathway inhibitor 2 (TFPI2), a Kunitz-type serine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service