Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6485

Sigma-Aldrich

GCSFR/CD114 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CSF3R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (N-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 69 kDa
observed mol wt 94 kDa by SDS-PAGE (reducing) (Glu 25 is the predicted N-terminal .)

packaging

pkg of 20 and 100 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GCSFR(1441)

General description

Granulocyte Colony Stimulating Factor Receptor (G-CSFR), also known as Cluster of Differentiation 114 (CD114), CSF3R and GCSF, is a cell-surface receptor for the granulocyte colony-stimulating factor (G-CSF), a cytokine that plays a critical role in the regulation of the activation, proliferation, differentiation, and survival of the neutrophilic granulocyte lineage. G-CSFR belongs to a family of cytokine receptors known as the hematopoietin receptor family. This type I membrane protein has a composite structure consisting of an immunoglobulin(Ig)-like domain, a cytokine receptor-homologous (CRH) domain and three fibronectin type I?II (FNIII) domains in the extracellular region. G-CSFR is present mainly on precursor cells in the bone marrow, and, in response to stimulation by G-CSF, initiates cell proliferation and differentiation into mature neutrophilic granulocytes and macrophages. G-CSFR mediates the specific effect of GCSF through activating a variety of intracellular signaling cascades, including the Jak/Stat, PI3/Akt, Ras-Raf-MAP kinase, and Src family kinase pathways, and thus functions in defense against infection, inflammation and repair, and in the maintenance of steady state hematopoiesis. Mutations in this gene are a cause of Kostmann syndrome, also known as severe congenital neutropenia. Mutations in the intracellular part of this receptor are also associated with certain types of leukemia.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service