Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6479

Sigma-Aldrich

FABP2/I-FABP human

recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE)

Synonym(s):

FABPI, I-FABP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (N-terminus)

assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 16 kDa
observed mol wt 16 kDa (DTT-reduced. Ala 2 is the predicted N-terminal .)

packaging

pkg of 10 and 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FABP2(2169)

General description

Fatty acid-binding protein 2 (FABP2), is also known as Fatty acid-binding protein, intestinal (FABPI), Intestinal-type fatty acid-binding protein (I-FABP). FABP2 belongs to the calycin superfamily and Fatty-acid binding protein (FABP) family. FABP2 / FABPI is expressed in the small intestine and at much lower levels in the large intestine and is highest expressed in the jejunum. FABP are thought to play a role in the intracellular transport of long-chain fatty acids and their acyl-CoA esters. FABP2 is probably involved in triglyceride-rich lipoprotein synthesis. FABP2 binds saturated long-chain fatty acids with a high affinity, but binds with a lower affinity to unsaturated long-chain fatty acids. FABP2 may also help maintain energy homeostasis by functioning as a lipid sensor.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service