Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6468

Sigma-Aldrich

R-Spondin3/RSPO3(22-146) human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CRISTIN1, PWTSR, THSD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 15.3 kDa
observed mol wt 18 kDa (DTT-reduced. Protein migrates due to glycosylation. Gln 22 is the predicted N-terminal.)
observed mol wt 24 kDa

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RSPO3(84870)

General description

R-Spondin 3 (RSPO3), also called cysteine-rich and single thrombospondin domain containing-1 (CRISTIN1), Protein with TSP type-1 repeat (PWTSR), is a member of the R-Spondin protein family. R-spondins (RSPO) are a recently discovered secretory protein family with four members in human and mouse. Although all four RSPO proteins activate the canonical Wnt pathway, RSPO2 and RSPO3 are more potent than RSPO1, whereas RSPO4 is relatively inactive. RSPO-3 is expressed ubiquitously and expressed at higher level in placenta, small intestine, fetal thymus and lymph node. RSPO3 is the activator of the β-catenin signaling cascade, leading to TCF-dependent gene activation. RSPO3 acts both in the canonical Wnt/β-catenin-dependent pathway and in non-canonical Wnt signaling pathway, probably by acting as an inhibitor of ZNRF3, an important regulator of the Wnt signaling pathway. RSPO3 also acts as a ligand for frizzled FZD8 and LRP6 and may negatively regulate the TGF-β pathway.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service