Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6463

Sigma-Aldrich

SFRP1/SARP2 human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Synonym(s):

FRP, FRP1, FrzA, SARP2, SFRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥92% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 33.4 kDa
observed mol wt 38 kDa (DTT-reduced. Protein migrates due to glycosylation. Ser 32 is the predicted N-terminal.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SFRP1(6422)

General description

Secreted frizzled-related protein 1 (SFRP1) also known as Secreted apoptosis-related protein 2 (SARP2), FRP, FRP1, is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. The Secreted frizzled-related protein (SFRP) family consists of five secreted glycoproteins in humans (SFRP1, SFRP2, SFRP3, SFRP4, SFRP5) that act as extracellular signaling ligands. SFRP1 / SARP-2 is widely expressed and absent from lung, liver and peripheral blood leukocytes, and with the highest levels in heart and fetal kidney. SARP2 / SFRP-1 is a 35 kDa prototypical member of the SFRP family. SARP2 / FRP-1 acts as a biphasic modulator of Wnt signaling, counteracting Wnt-induced effects at high concentrations and promoting them at lower concentrations.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service