Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6460

Sigma-Aldrich

CD147 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

5F7, BSG, Basigin, EMMPRIN, M6, OK, TCSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 21.6 kDa
observed mol wt 26-33 kDa (DTT-reduced. Protein migrates due to glycosylation. Ala 22 is the predicted N-terminal.)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BSG(682)

General description

CD147, also known as Basigin (BSG), or extracellular matrix metalloproteinase inducer (EMMPRIN). The human basigin protein contains 269 amino acids that form two heavily glycosylated C2 type immunoglobulin-like domains at the N-terminal extracellular portion. A second form of basigin has also been characterized that contains one additional immunoglobulin-like domain in its extracellular portion. As members of the immunoglobulin superfamily play fundamental roles in intercellular recognition involved in various immunologic phenomena, differentiation, and development, basigin is thought also to play a role in intercellular recognition and regulate several distinct functions, such as spermatogenesis, expression of the monocarboxylate transporter and the responsiveness of lymphocytes. Basigin is a type I integral membrane receptor that has many ligands, including the cyclophilin (CyP) proteins Cyp-A and CyP-B and certain integrins.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aimei Lee et al.
Journal of gastroenterology and hepatology, 31(2), 459-466 (2015-08-28)
The glycoprotein CD147 has a role in tumor progression, is readily detectable in the circulation, and is abundantly expressed in hepatocellular carcinoma (HCC). Advanced HCC patients are a heterogeneous group with some individuals having dismal survival. The aim of this
David Heinzmann et al.
Biomolecules, 10(4) (2020-04-08)
Surface receptor-mediated adhesion is a fundamental step in the recruitment of leukocytes and platelets, as well as platelet-leukocyte interactions. The surface receptor CD147 is crucially involved in host defense against self-derived and invading targets, as well as in thrombosis. In
Michal A Rahat et al.
Arthritis research & therapy, 22(1), 240-240 (2020-10-16)
Angiogenesis plays a central role in the pathophysiology of rheumatic diseases. Patients with psoriatic arthritis (PsA) demonstrate increased vascularity over patients with rheumatoid arthritis (RA), with unknown mechanisms. We evaluated the serum levels of several pro- and anti-angiogenic factors in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service