Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6452

Sigma-Aldrich

IL20RB/IL-20R2 human

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)

Synonym(s):

DIRS1, FNDC6, IL-20R-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 49.1 kDa (This protein fused with Fc fragment of human IgG1 at the C-terminus.)
observed mol wt 50 kDa (DTT reduced. 100 kDa non reduced. Asp 30 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... IL20RB(53833)

General description

Interleukin-20 receptor subunit β (IL20RB) is also known as IL-20R2, DIRS1, which belongs to the type II cytokine receptor family. IL20RB / IL-20R2 contains two fibronectin type-III domains. IL20RB / IL-20R2 is widely expressed with highest levels in skin and testis. IL20RB can be fused with IL20RA or IL22RA1 forming heterodimer. The IL20RA/IL20RB dimer is a receptor for IL19, IL20 and IL24. The IL22RA1/IL20RB dimer is a receptor for IL20 and IL24.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 100 mM glycine, pH 7.0. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service