Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6449

Sigma-Aldrich

LIFR/CD118 human

recombinant, expressed in HEK 293 cells, ≥96% (SDS-PAGE)

Synonym(s):

Cluster of Differentiation 118, FLJ98106, FLJ99923, LIF-R, Leukemia inhibitory factor receptor, SJS2, STWS, SWS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in HEK 293 cells

assay

≥96% (SDS-PAGE)

form

lyophilized

mol wt

monomer calculated mol wt 115.5 kDa (This protein is fused with the Fc region of human IgG1 at the CT.)
observed mol wt 135-150 kDa by SDS-PAGE (reducing)

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LIFR(3977)

General description

Leukemia inhibitory factor receptor also known as LIFR; CD118; FLJ98106; FLJ99923; LIF-R; SJS2; STWS; SWS, is the receptor for leukemia inhibitory factor (LIF). The leukemia inhibitory factor is a polyfunctional cytokine that affects the differentiation, survival, and proliferation of a wide variety of cells in the adult and the embryo. LIF action appears to be mediated through a high-affinity receptor complex composed of a low-affinity LIF binding chain (LIF receptor) and a high-affinity converter subunit, gp130. Both LIFR and gp130 are members of a family of cytokine receptors that includes components of the receptors for the majority of hematopoietic cytokines and for cytokines that affect other systems, including the ciliary neurotrophic factor, growth hormone and prolactin Defects in LIFR are the cause of Stueve-Wiedemann syndrome (SWS), a severe autosomal recessive condition and belongs to the group of the bent-bone dysplasias.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service