Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP6448

Sigma-Aldrich

Fas Ligand/FasL human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

ALPS1B, APT1LG1, CD178, CD95-L, CD95L, FASL, FASLG, Fas ligand, TNFSF6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (N-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 17.7 kDa
observed mol wt 25-32 kDa by SDS-PAGE (reducing) (His is the predicted N-terminal .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FASLG(356)

General description

Fas ligand also known as FasL, CD178, CD95L, or TNFSF6, is a 40 kDa homotrimeric type-II transmembrane protein that belongs to the tumor necrosis factor (TNF) family.[1] The FASL gene was mapped to human chromosome 1q24.3.[2] It spans a length of 8 kb and contains 4 exons.[3] It is found to be expressed on T and NK cells that are activated.[4]

Biochem/physiol Actions

The binding of Fas ligand to its receptor induces apoptosis. Fas ligand/receptor interactions play an important role in the regulation of the immune system and the progression of cancer. Its functions include T-cell homeostasis, cytotoxic T-cell activity, immune privilege, maternal tolerance, tumor counterattack.[1] Defective Fas-mediated apoptosis may lead to oncogenesis as well as drug resistance in existing tumors. Germline mutation of Fas is associated with autoimmune lympho proliferative syndrome (ALPS), a childhood disorder of apoptosis.[5]

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 100 mM glycine, pH 7.0. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominant interfering Fas gene mutations impair apoptosis in a human autoimmune lymphoproliferative syndrome.
Fisher G H, et al.
Cell, 81(6), 935-946 (1995)
Insights into the molecular regulation of FasL (CD178) biology.
Lettau M, et al.
European Journal of Cell Biology, 90(6-7), 456-466 (2011)
Metalloproteinase-mediated release of human Fas ligand.
Kayagaki N, et al.
The Journal of Experimental Medicine, 182(6), 1777-1783 (1995)
Statistical colocalization of genetic risk variants for related autoimmune diseases in the context of common controls.
Fortune M D, et al.
Nature Genetics, 47(7), 839-839 (2015)
T Takahashi et al.
International immunology, 6(10), 1567-1574 (1994-10-01)
Fas ligand (FasL) is a 40 kDa type II membrane protein belonging to the tumor necrosis factor family, which induces apoptosis by binding to its receptor, Fas. In this report, we isolated the chromosomal gene for human FasL. The human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service