Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6444

Sigma-Aldrich

SEMA4D/CD100 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

C9orf164, FLJ33485, FLJ34282, FLJ39737, FLJ46484, M-sema-G, MGC169138, MGC169141, SEMAJ, Semaphorin-4D, coll-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 105.4 kDa (This protein is fused with Fc fragment of human IgG1 at the C-terminus.)
observed mol wt 120-140 kDa by SDS-PAGE (reducing) (Met 22 is the predicted N-terminal .)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SEMA4D(10507)

General description

Cluster of Differentiation 100 (CD100), also known as Semaphorin-4D (SEMA4D), is a single-pass type I membrane protein which belongs to the semaphorin family, and is a human 150-kDa homodimer expressed at the surface of most hemopoietic cells. Semaphorin genes encode soluble and membrane-bound proteins, most of which have been shown to act as chemorepellents on growth cone guidance. CD100 is discrete, as it is a transmembrane leukocyte surface molecule that can also exist in a soluble form. Semaphorin 4D (Sema 4D) is an axon guidance molecule which is secreted by oligodendrocytes and induces growth cone collapse in the central nervous system. By binding plexin B1 receptor it functions as an R-Ras GTPase-activating protein (GAP) and repels axon growth cones in both the mature central nervous system. In the immune system, CD100 binds CD72 to activate B cells and dendritic cells, though much about this interaction is still under investigation. It is involved in oligodendrogenesis during development and during recovery from ischemic injury.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 100 mM glycine, pH 7.0. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service