Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6442

Sigma-Aldrich

CD166/ALCAM human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CD6L, KG-CAM, MEMD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 56.8 kDa
observed mol wt 65-80 kDa (DTT-reduced. Trp28 is the predicted N-terminal .)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ALCAM(214)

General description

CD166 antigen is a 100-105 kDa type I transmembrane glycoprotein that is a member of the immunoglobulin superfamily of proteins, which is also known as activated leukocyte cell adhesion molecule (ALCAM) in human, SC-1/DM-GRASP/BEN in the chicken, and KG-CAM in the rat. CD166 is a cell adhesion molecule that binds to CD6. CD166 involved in neurite extension by neurons via heterophilic and homophilic interactions.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service