Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6433

Sigma-Aldrich

PD-L1/CD274/B7-H1 from mouse

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

PDCD1L1, PDCD1LG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 25.6 kDa
observed mol wt 40-50 kDa (DTT-reduced. Protein migrates due to glycosylation. Phe 19 is the predicted N-terminal.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg (LAL test)

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

mouse ... CD274(60533)

General description

Programmed cell death 1 ligand 1 (PD-L1), which is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules. PD-L1 is a cell surface immunoglobulin with two Ig-like domains within the extracellular region and a short cytoplasmic domain. The protein is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes.[1][2][3]

Biochem/physiol Actions

Programmed cell death 1 ligand 1 (PD-L1) is involved in the regulation of cellular and humoral immune responses. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after lipopolysaccharide (LPS) and interferon-γ (IFNG) activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 is involved in the co-stimulatory signal, essential for T-cell proliferation and production of interleukin-10 (IL10) and IFNG, in an interleukin-2-dependent and a programmed cell death 1 (PDCD1)-independent manner.[4]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intrinsic and extrinsic control of expression of the immunoregulatory molecule PD-L1 in epithelial cells and squamous cell carcinoma.
Ritprajak P and Azuma M
Oral Oncology, 51(3), 221-228 (2015)
Integrative analysis reveals selective 9p24.1 amplification, increased PD-1 ligand expression, and further induction via JAK2 in nodular sclerosing Hodgkin lymphoma and primary mediastinal large B-cell lymphoma.
Green MR
Blood, 116(17), 3268-3277 (2010)
The role of B7 family molecules in hematologic malignancy.
Greaves P and Gribben JG
Blood, 121(5), 734-744 (2013)
Adnan R Khan et al.
Nature communications, 6, 5997-5997 (2015-01-23)
Specific B-cell subsets can regulate T-cell immune responses, and are termed regulatory B cells (Breg). The majority of Breg cells described in mouse and man have been identified by IL-10 production and are known to suppress allergy and autoimmunity. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service