Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6424

Sigma-Aldrich

SERPIN F1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

EPC-1, PEDF, PIG35, Serpin F1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 45.2 kDa
observed mol wt 45-55 kDa (DTT-reduced. Protein migrates due to glycosylation. Gln 20 is the predicted N-terminal.)

packaging

pkg of 10 μg
pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPINF1(5176)

General description

Serpin F1 (SERPINF1) is also known as Pigment epithelium-derived factor (PEDF), Cell proliferation-inducing gene 35 protein (PIG35). Serpin F1 belongs to the serpin family. Serpin F1 is expressed in quiescent cells. PEDF has a variety of functions including antiangiogenic, antitumorigenic, and neurotrophic properties. Endothelial cell migration is inhibited by SERPINF1/ PEDF. PEDF / SERPINF1 suppresses retinal neovascularization and endothelial cell proliferation. PEDF is also responsible for apoptosis of endothelial cells either through the p38 MAPK pathway or through the FAS/FASL pathway. PEDF also displays neurotrophic functions.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7.4. Normally Mannitol or Trehalose are added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service