Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SRP6389

Sigma-Aldrich

TNFR1/CD120a/TNFRSF1A human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Synonym(s):

FPF, TBP1, TNF-R, TNF-R-I, TNF-R55, TNFAR, TNFR55, TNFR60, p55, p60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥92% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 22 kDa
observed mol wt 30-35 kDa (DTT-reduced. Protein migrates due to different glycosylation. Ile 22 is the predicted N-terminus.)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF1A(7132)

General description

Tumor necrosis factor receptor 1 (TNF-R1) also known as Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A), TNFAR, CD antigen CD120a, belongs to the tumor necrosis factor receptor superfamily. TNF-R1 contains one death domain and four TNFR-Cys repeats. TNF-R1 is the receptor of TNFSF2 / TNF-α and homotrimeric TNFSF1 / lymphotoxin - α. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. TNF-R1 contributes to the induction of non - cytocidal TNF effects including anti-viral state and activation of the acid sphingomyelinase. Defects in TNFRSF1A are the cause of familial Hibernian fever (FHF).

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service