Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6386

Sigma-Aldrich

CD111/Nectin-1/PVRL1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CLPED1, ED4, HIgR, HVEC, OFC7, PRR, PRR1, PVRR, PVRR1, SK-12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 34.8 kDa
observed mol wt 45-60 kDa (DTT-reduced. Protein migrates due to glycosylation. Gln 31 is the predicted N-terminus.)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PVRL1(5818)

General description

Poliovirus receptor-related protein 1 (PVRL1) is also known as Herpes virus entry mediator C (HveC), Herpesvirus Ig-like receptor (HIgR), Nectin-1, CD antigen CD111, PVRL1 / Nectin-1 / CD111 belongs to the nectin family. PVRL1 / Nectin-1 is a membrane protein with three extracellular immunoglobulin domains, a single transmembrane helix and a cytoplasmic tail. The protein can mediate Ca2+-independent cellular adhesion further characterizing it as IgSF cell adhesion molecule (IgSF CAM). PVRL1 is adhesion molecule found in a wide range of tissues. PVRL1 / Nectin-1 promote cell-cell contacts by forming homophilic or heterophilic trans-dimers. Heterophilic interactions have been detected between PVRL1/nectin-1 and PVRL3/nectin-3 and between PVRL1/nectin-1 and PVRL4/nectin-4.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service