Urokinase plasminogen activator surface receptor (U-PAR) is also known as PLAUR, Monocyte activation antigen Mo3, CD antigen CD87. PLAUR contains three UPAR/Ly6 domains. U-PAR is expressed in neurons of the rolandic area of the brain (at protein level) and is also expressed in the brain. PLAUR / CD87 interacts with MRC2, SRPX2 and SORL1. PLAUR / UPAR act as a receptor for urokinase plasminogen activator and plays a role in localizing and promoting plasmin formation. U-PAR mediates the proteolysis-independent signal transduction activation effects of U-PA.
Physical form
Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.