SRP6378
CD36 human
recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Synonym(s):
GP3B, GP4, Platelet Glycoprotein 4, SCARB3
About This Item
Recommended Products
biological source
human
recombinant
expressed in HEK 293 cells
tag
6-His tagged (C-terminus)
assay
≥95% (SDS-PAGE)
form
lyophilized
mol wt
calculated mol wt 47.5 kDa
observed mol wt 60-90 kDa by SDS-PAGE (reducing) (Gly30 is the predicted N-terminus.)
packaging
pkg of 10 μg
pkg of 50 μg
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CD36(948)
General description
Physical form
Reconstitution
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service