Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6376

Sigma-Aldrich

CD273 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Butyrophilin B7-DC, PDCD1 ligand 2, PDCD1L2, PDCD1LG2, PDL2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 23.4 kDa
observed mol wt 35-48 kDa (DTT-reduced. Protein migrates due to glycosylation. Leu 20 is the predicted N-terminus.)

packaging

pkg of 10 μg
pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Programmed cell death 1 ligand 2 (PD-L2 or PDCD1 ligand 2) is also known as butyrophilin B7-DC or CD antigen CD273. It belongs to the immunoglobulin superfamily or BTN/MOG family. The gene encoding CD273 is localized on human chromosome 9p24.1.[1]

Biochem/physiol Actions

The expression of programmed cell death 1 ligand 2 (PD-L2) or CD antigen 273 (CD273) is up-regulated by interferon-γ (IFNG/IFN-γ) stimulation in monocytes and induced on dendritic cells grown from peripheral blood mononuclear cells with colony-stimulating factor-2 (CSF2) and interleukin-4 (IL-4).[1] PD-L2 is involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner.[2] PD-L2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.[3]

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PD-L2:PD-1 involvement in T cell proliferation,
cytokine production, and integrin-mediated adhesion
Paul. A. Saunders
European Journal of Immunology, 35 (2005)
No evidence for dualism in function and receptors: PD-L2/B7-DC is an inhibitory regulator of human T cell activation
Katrina P
European Journal of Immunology, 36(5), 1104-1113 (2006)
Expression of Programmed Cell Death 1 Ligand 2 (PD-L2) is a Distinguishing Feature of Primary Mediastinal (Thymic) Large B-cell Lymphoma and Associated with PDCD1LG2 Copy Gain
Min Shi
American Journal of Surgical Pathology, 38(12), 1715-1723 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service