Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6365

Sigma-Aldrich

KLRC3 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Killer cell lectin-like receptor subfamily C, NKG2-E, NKG2E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

19 kDa (171 aa, 94-240 aa + His Tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KLRC3(3823)

General description

KLRC3 is a member of the NKG2 group which are expressed primarily in natural killer (NK) cells and encodes a family of transmembrane proteins characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed on NK cells. Recombinant human KLRC3 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M NaCl and 10% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service