Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6364

Sigma-Aldrich

KLRC2 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

CD159c, NKG2-C, NKG2-C type II integral membrane protein, NKG2C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥85% (SDS-PAGE)

form

liquid

mol wt

18.4 kDa (162 aa, 94-231 aa + His Tag)

packaging

pkg of 50 μg

concentration

0.5 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KLRC2(3822)

General description

NKG2-C type II integral membrane protein, also known as KLRC2, plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. The group, designated KLRC (NKG2) are expressed primarily in natural killer (NK) cells and encodes a family of transmembrane proteins characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The KLRC (NKG2) gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed on NK cells. KLRC2 alternative splice variants have been described but their full-length nature has not been determined. Recombinant human KLRC2 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

0.5 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M Urea and 10% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service