Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6355

Sigma-Aldrich

CD68 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

GP110, LAMP4, Macrosialin, SCARD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥85% (SDS-PAGE)

form

liquid

mol wt

34.1 kDa (323 aa, 22-319 aa + His Tag)

packaging

pkg of 50 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD68(968)

General description

Macrosialin, also known as CD68, is a member of the lysosomal/endosomal-associated membrane glycoprotein (LAMP) family. It is found in cytoplasmic granules and in the cytoplasm of various non-hematopoietic tissues including liver and kidney tubules and glomeruli. CD68 is also found, to a lesser extent, on the surface of macrophages, monocytes, neutrophils, basophils and large lymphocytes. Recombinant human CD68 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.1 M NaCl, 1 M Urea and 10% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service