Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6351

Sigma-Aldrich

CD244 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

2B4, CD244 molecule, NAIL, NKR2B4, Nmrk, SLAMF4, natural killer cell receptor 2B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥85% (SDS-PAGE)

form

liquid

mol wt

25.5 kDa (230 aa, 19-224 aa + His Tag)

packaging

pkg of 50 μg

concentration

0.5 mg/mL

UniProt accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

human ... CD244(51744)

General description

CD244 (Natural Killer Cell Receptor 2B4), also known as Cluster of Differentiation 244, contains 2 Ig-like (immunoglobulin-like) domains. A role for the subtypes of CD2 Ig superfamily receptors has been recently demonstrated in eosinophilic inflammation in experimental asthma and atopic asthmatics. Functions of CD244 molecules are in eosinophil adhesion and chemotaxis, and correlated the results to the pathophysiology of allergic rhinitis (AR). The cluster of differentiation (cluster of designation) (often abbreviated as CD) is a protocol used for the identification and investigation of cell surface molecules present on white blood cells initially but found in almost any kind of cell of the body, providing targets for immunophenotyping of cells. Recombinant human CD244 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

0.5 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M Urea. .

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service