This product is suitable for Western blot applications and not mammalian cell culture applications.
SRP6348
ULBP2 human
recombinant, expressed in E. coli, ≥85% (SDS-PAGE)
Synonym(s):
N2DL2, NKG2D ligand 2, RAET1H
About This Item
biological source
human
recombinant
expressed in E. coli
assay
≥85% (SDS-PAGE)
form
liquid
mol wt
24.3 kDa (216 aa, 26-216 aa + His Tag)
packaging
pkg of 100 μg
concentration
1 mg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... ULBP2(80328)
General description
Physical form
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is this recombinant protein suitable for stimulations in mammalian cell culture (e.g.: PBMCs)?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service