Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP6345

Sigma-Aldrich

TNFSF8 human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

CD153, CD30L, CD30LG, tumor necrosis factor ligand superfamily member 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

liquid

mol wt

22 kDa (195 aa, 63-234 aa + His Tag)

packaging

pkg of 50 μg

concentration

0.5 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TNFSF8(944)

General description

TNFSF8 is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF8/CD30, which is a cell surface antigen and a marker for Hodgkin lymphoma and related hematologic malignancies. The engagement of this cytokine expressed on B cell surface plays an inhibitory role in modulating Ig class switch. TNFSF8 was shown to enhance cell proliferation of some lymphoma cell lines, while to induce cell death and reduce cell proliferation of other lymphoma cell lines. The pleiotropic biologic activities of this cytokine on different CD30+ lymphoma cell lines may play a pathophysiologic role in Hodgkin′s and some non-Hodgkin′s lymphomas. Recombinant human TNFSF8 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

0.5 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol and 0.4 M Urea.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service