Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6343

Sigma-Aldrich

SFTPD human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

COLEC7, PSP-D, Pulmonary surfactant-associated protein D precursor, SFTP4, SP-D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥80% (SDS-PAGE)

form

liquid

mol wt

18.9 kDa (175 aa, 224-375 aa + His Tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SFTPD(6441)

General description

SFTPD (surfactant protein D) is a Ca2+-binding, large multimeric hydrophilic protein, which is produced by type II pneumocytes and club cells. It lines the alveolar epithelium. It is a member of collectins (collagen-containing C-type lectins) which are involved in innate responses.
SFTPD, as known as Surfactant pulmonary-associated protein D, is a member of the collectin family of C-type lectins that is synthesized in many tissues including respiratory epithelial cells in the lung, and contains one C-type lectin domain and one collagen-like domain. It is humoral molecules of the innate immune system, and is considered a functional candidate in chronic periodontitis. It is also involved in the development of acute and chronic inflammation of the lung. Several human lung diseases are characterized by decreased levels of bronchoalveolar SFTPD. Recombinant human SFTPD protein, fused to His-tag at N-terminus, was expressed in E.coli.
SFTPD (surfactant protein D) encodes an extracellular soluble protein that is expressed in the mucosal membrane. The SFTPD gene is mapped to human chromosome 10q22.3.

Biochem/physiol Actions

SFTPD (surfactant protein D) protein is thought to decrease inflammatory responses in alveolar macrophages and oxidant production, and increase apoptotic cell clearance. Elevated serum levels of this protein function as biomarker for susceptibility to COPD (chronic obstructive pulmonary disease). It is involved in the pathogenesis of COPD, and certain polymorphisms in this gene are linked with changes in its serum level and lung function. In human bronchoalveolar lavage, SFTPD is the predominant protein involved in antiviral response against seasonal strains of influenza A virus (IAV). Studies in mice show that the neck and carbohydrate recognition domain (NCRD) of this protein confers protection against allergy and respiratory syncytial virus infection. Variations in this gene are linked with lung function measures in context of tobacco smoking. Hence, this protein has potential as a biomarker for subclinical tobacco smoke-induced lung damage.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, and 0.4 M Urea.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tremor?ataxia with central hypomyelination (TACH) leukodystrophy maps to chromosome 10q22. 3?10q23. 31.
Bernard G, et al.
Neurogenetics, 11(4), 457-464 (2010)
Ryuta Saka et al.
Pediatric surgery international, 32(1), 59-63 (2015-10-30)
Necrotizing enterocolitis (NEC) is a devastating inflammatory disease of preterm infants that may depend on overexpression of toll-like receptor-4 (TLR4) in the immature intestine. Surfactant protein (SP)-D is a member of the collectin family and plays an important role in
Kun Cao et al.
Journal of inorganic biochemistry, 182, 113-123 (2018-02-20)
Iron is an essential element for almost all bacteria. The iron ATP-binding cassette (ABC) transporters located on the cell membrane affects bacterial virulence and infection. Although a variety of Fe3+-transporters have been found in bacteria, their evolutionary processes are rarely
Abdullah Kumral et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 30(4), 446-451 (2016-04-26)
Periventricular leukomalacia (PVL) is the leading cause of neurocognitive deficits in children with prematurity. We previously hypothesized that surfactant protein D (SPD) with its ability to bind toll-like receptors may have a possible ameliorating effect in PVL. Three groups were
Christian A Ruge et al.
PloS one, 7(7), e40775-e40775 (2012-07-18)
The peripheral lungs are a potential entrance portal for nanoparticles into the human body due to their large surface area. The fact that nanoparticles can be deposited in the alveolar region of the lungs is of interest for pulmonary drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service