Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6338

Sigma-Aldrich

IRF2 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

Interferon regulatory factor-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

15 kDa (133 aa, 1-113 aa + His tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IRF2(3660)

General description

Interferon regulatory factor (IRF) 2 is a member of the interferon regulatory transcription factor family. IRF-2 is generally regarded as an oncoprotein. Structure of IRF1 and IRF2 are similar. However, its functions are different. IRF2 competitively inhibits the IRF1-mediated transcriptional activation of interferons α and β, and presumably other genes that employ IRF1 for transcription activation. Recombinant human IRF2 was expressed in E.coli and purified by using conventional chromatography techniques.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0), 1 mM DTT and 10% glycerol.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide analysis of cis-regulatory changes underlying metabolic adaptation of cavefish.
Krishnan, et al.
Nature Genetics, 54, 684-693 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service