Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6337

Sigma-Aldrich

IRF1 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

Interferon regulatory factor-1, MAR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

15 kDa (134 aa, 1-114 aa + His tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IRF1(3659)

General description

IRF1, Interferon regulatory factor 1, is a member of the interferon regulatory transcription factor (IRF) family which regulates gene expression critical to immune response, hematopoiesis and proliferation. IRF-1 is a transcriptional activator for IFN-α, IFN-β, and IFN-gamma stimulated genes. IRF1 is also a tumor suppressor transcription factor inducing apoptosis of tumorigenic cell lines. Recombinant human IRF1 was expressed in E.coli and purified by using conventional chromatography techniques.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) and 10% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth J Dery et al.
Molecular cancer, 13, 64-64 (2014-03-22)
Interferon regulatory factor-1 (IRF-1) is a master regulator of IFN-γ induced gene transcription. Previously we have shown that IRF-1 transcriptionally induces CEACAM1 via an ISRE (Interferon-Stimulated Response Element) in its promoter. CEACAM1 pre-mRNA undergoes extensive alternative splicing (AS) generating isoforms
Tomoyuki Ohsugi et al.
Oncogene, 38(32), 6051-6064 (2019-07-12)
Impaired Wnt signaling pathway plays a crucial role in the development of colorectal cancer through activation of the β-catenin/TCF7L2 complex. Although genes upregulated by Wnt/β-catenin signaling have been intensively studied, the roles of downregulated genes are poorly understood. Previously, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service