SRP6331
CD5 human
recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Synonym(s):
LEU1, T-cell surface glycoprotein CD5, T1
About This Item
biological source
human
recombinant
expressed in E. coli
assay
≥80% (SDS-PAGE)
form
liquid
mol wt
41 kDa (371 aa, 25-372 aa + His tag)
packaging
pkg of 50 μg
concentration
1 mg/mL
impurities
<1 EU/μg endotoxin (LAL test)
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CD5(921)
General description
Recombinant human CD5 protein, fused to His-tag at N-terminus, was expressed in Escherichia coli.
Biochem/physiol Actions
Physical form
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service