Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6325

Sigma-Aldrich

CD200 human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

CD200 molecule, MOX1, MOX2, MRC, OX-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

24.8 kDa (225 aa, 31-232 aa + His tag)

packaging

pkg of 50 μg

concentration

1 mg/mL

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD200(4345)

General description

CD200 is a type-1 membrane glycoprotein, which contains two immunoglobulin domains, and thus belongs to the immunoglobulin superfamily. Studies of the related genes in mouse and rat suggest that this gene may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. Recombinant human CD200 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4M Urea.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanislava Pankratova et al.
Molecular neurobiology, 53(1), 584-594 (2014-12-17)
The CD200 ligand is expressed by a variety of cell types, including vascular endothelia, kidney glomeruli, some subsets of T and B cells, and neurons in the brain and periphery. In contrast, the receptor of CD200, CD200R, has a limited
Li-Chun Weng et al.
Journal of reproductive immunology, 138, 103090-103090 (2020-02-06)
Endometriosis (EMS) is a benign disease that is related to estrogen, immune disorders and inflammation. The purpose of this research was to determine the expression of CD200 in EMS and to clarify its role in the pathogenesis of the disease.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service