Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6294

Sigma-Aldrich

IL-7 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Interleukin-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 17 kDa (no tag)
observed mol wt 19 kDa (DTT-reduced. Protein migrates due to glycosylation. Asp26 is the predicted N-terminus.)
observed mol wt 22 kDa
observed mol wt 26 kDa

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL-7(3574)

General description

Interleukin 7, also known as IL7, IL-7, and is a hematopoietic growth factor secreted by stromal cells in the red marrow and thymus. It is also produced by keratinocytes, dendritic cells, hepatocytes, neurons, and epithelial cells but is not produced by lymphocytes. IL-7 stimulates the differentiation of multipotent (pluripotent) hematopoietic stem cells into lymphoid progenitor cells. It also stimulates proliferation of all cells in the lymphoid lineage (B cells, T cells and NK cells). It is important for proliferation during certain stages of B-cell maturation, T and NK cell survival, development and homeostasis. IL-7 is a cytokine important for B and T cell development. This cytokine and the hepatocyte growth factor (HGF) form a heterodimer that functions as a pre-pro-B cell growth-stimulating factor. IL-7 binds to the IL-7 receptor, a heterodimer consisting of Interleukin-7 receptor α and common gamma chain receptor. Il-7 promotes hematological malignacies (acute lymphoblastic leukemia, T cell lymphoma). Elevated levels of IL-7 have also been detected in the plasma of HIV-infected patients. IL-7 as an immunotherapy agent has been examined in many pre-clinical animal studies and more recently in human clinical trials for various malignancies and during HIV infection. IL-7 could also be beneficial in improving immune recovery after allogenic stem cell transplant.
Interleukin-7 (IL-7) is a cytokine. It is synthesized by stromal cells, epithelial cells, endothelial and immune cells.[1][2] The IL-7 gene is located on human chromosome 8q21.[3]

Biochem/physiol Actions

Interleukin-7 (IL-7) plays a major role in innate and adaptive immunity.[1] It participates in B and T cell development and osteoblasts differentiation of multiple myeloma.[2] IL-7 is also involved in the development, survival and proliferation of lymphocytes.[3] It has the ability to prevent the initiation of MAPK signaling.[2]

Physical form

Lyophilized from 0.22 μm filtered solution in in PBS, pH 7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile deionized water to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elevated systemic interleukin-7 in patients with colorectal cancer and individuals at high risk of cancer: association with lymph node involvement and tumor location in the right colon.
Krzystek-Korpacka M, et al.
Cancer Immunology, Immunotherapy, 66(2), 171-179 (2017)
IL-7 suppresses osteogenic differentiation of periodontal ligament stem cells through inactivation of mitogen-activated protein kinase pathway.
Jian CX, et al.
Organogenesis, 12(4), 183-193 (2016)
Overexpression of IL-7 enhances cisplatin resistance in glioma.
Cui L, et al.
Cancer Biology & Therapy, 13(7), 496-503 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service