Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6289

Sigma-Aldrich

GFER/HPO/ALR human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Synonym(s):

ERV1, HERV1, HPO1, HPO2, HSS, Hepatopoietin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥92% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 15.2 kDa (no tag)
observed mol wt 16.0 kDa (DTT-reduced. Met 81 is the predicted N-terminus .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GFER(2671)

General description

Growth factor, augmenter of liver regeneration (GFER) is also known as FAD-linked sulfhydryl oxidase ALR, which belongs to the Erv1/ALR family of proteins. This family can be found in higher and lower eukaryotes.[1] There are two isoform of GFER: Isoform 1 and isoform 2. Isoform 2 missing 1 – 80 aa. Isoform 1 is mainly located in mitochondrial intermembrane space, while Isoform 2 is secreted to cytoplasm.

Biochem/physiol Actions

Growth factor, augmenter of liver regeneration (GFER) controls mitochondrial fission and apoptosis in liver ischemia.[2] Isoform 1 of GFER regenerates the redox-active disulfide bonds in CHCHD4/MIA40, a chaperone essential for disulfide bond formation and protein folding in the mitochondrial intermembrane space. The reduced form of coiled-coil-helix-coiled-coil-helix domain containing 4 (CHCHD4/MIA40) forms a transient intermolecular disulfide bridge with GFER/ERV1 (sulfhydryl oxidase), resulting in regeneration of the essential disulfide bonds in CHCHD4/MIA40, while GFER/ERV1 becomes re-oxidized by donating electrons to cytochrome c or molecular oxygen. The isoform 2 of GFER may act as an autocrine hepatotropic growth factor promoting liver regeneration.

Physical form

Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 100 mM NaCl, pH 8.0. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 100 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Augmenter of liver regeneration.
Gandhi C R
Fibrogenesis & Tissue Repair, 5(1), 10-10 (2012)
Hepatic stimulator substance resists hepatic ischemia/reperfusion injury by regulating Drp1 translocation and activation.
Zhang C, et al.
Hepatology, 66(6), 1989-2001 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service