Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6286

Sigma-Aldrich

PCSK9 murine

recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE)

Synonym(s):

FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9, PCSK9, Proprotein convertase subtilisin/kexin type 9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in HEK 293 cells

tag

6-His tagged (N-terminus)

assay

≥97% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 72 kDa
observed mol wt 20 kDa (DTT-reduced. Protein migrates due to glycosylation. Gln 35 is the predicted N-terminus .)
observed mol wt 64 kDa

packaging

pkg of 20 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

mouse ... PCSK9(100102)

General description

Proprotein convertase subtilisin/kexin type 9 (PCSK9), is an enzyme which in humans is encoded by the PCSK9 gene. This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein plays a major regulatory role in cholesterol homeostasis. PCSK9 binds to the epidermal growth factor-like repeat A (EGF-A) domain of the low-density lipoprotein receptor (LDLR), inducing LDLR degradation. PCSK9 may also have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a rare form of autosomal dominant familial hypercholesterolemia (HCHOLA3).

Physical form

Sterile filtered through a 0.22 micron filter. Lyophilized from 1 x PBS, pH 7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 100 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service