Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6280

Sigma-Aldrich

BRD3(306-417) his tag

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

KIAA0043, RING3L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

liquid

mol wt

15.5 kDa

packaging

pkg of 1 mg
pkg of 100 μg
pkg of 20 μg

concentration

2 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRD3(8019)

General description

The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. BRD3 binds hyper-acetylated chromatin and plays a role in the regulation of transcription, probably by chromatin remodeling and interaction with transcription factors. It regulates transcription by promoting the binding of the transcription factor GATA1 to its targets and transcription of the CCND1 gene. A chromosomal aberration involving BRD3 was found in a rare, aggressive, and lethal carcinoma arising in midline organs of young people. The recombinant protein includes Bromodomain-containing protein 3 (contains 306-417 aa) with N-terminal His-tag.

Physical form

rh-BRD3-BD2 is supplied as a solution in PBS Buffer containing 10% glycerol.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service