Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6240

Sigma-Aldrich

TWEAK Receptor human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

FGFinducible 14, TNF-related weak inducer of apoptosis receptor, TNFRSF12A, TWEAKR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

5.6 kDa

packaging

pkg of 10 μg
pkg of 50 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

TWEAKR belongs to the TNFR superfamily of transmembrane proteins and contains a cytoplasmic “death domain,” which can activated the cell′s apoptotic machinery. It is expressed in the spleen, thymus, peripheral blood lymphocytes, colon, and small intestine. Signal transduction by TWEAKR can be activated by either the membrane anchored or the soluble TWEAK. Recombinant soluble TWEAKR is a 53 amino acid polypeptide (5.6 kDa) comprising the entire extracellular domain of the full length TWEAKR protein.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 10 mM Sodium Phosphate, pH 7.5 and 50 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service