Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6239

Sigma-Aldrich

TSG human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

Twisted Gastrulation Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

22.2 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TSG(57045)

General description

Twisted Gastrulation Protein (TSG) is a secreted BMP binding protein structurally related to the BMP antagonists Chordin and Noggin. TSG can inhibit BMP activity by binding directly to BMP proteins, and can act either as a BMP4 agonist or antagonist (depending on the specific biochemical environment) by binding to the BMP4/Chordin complex. Recombinant human TSG is a 198 amino acid 22.2 kDa protein containing the BMP/TGFβ binding portion of the full length TSG protein.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 10 mM Sodium Acetate, pH 6.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service