Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6235

Sigma-Aldrich

SDF-1α/CXCL2 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

CXCL12, PBSF, Stromal-Cell Derived Factor-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

7.9 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

SDF-1 (stromal-derived factor 1) is a constitutive gene which is ubiquitously expressed in vertebrate tissues. It was first identified in rat bone marrow (BM) stromal cell line supporting hematopoietic cell culture. This gene is regulated at splicing level, rather than transcriptional level and is spliced into multiple variants. Among these, SDF-1α is the major isoform expressed in all tissues, but is rapidly proteolysed in blood. This isoform is composed of 89 amino acids. SDF-1 is a member of the intercrine cytokine family or chemokine family of proteins, which are characterized by four conserved cysteines that form two disulfide bonds.
Recombinant rat SDF-1α is a 7.9kDa protein containing 68 amino acid residues.

Biochem/physiol Actions

SDF-1 (stromal-derived factor 1) performs a key function in cell migration and proliferation and in tissue-specific processes such as neuromodulation. This gene is also involved in chronic inflammatory disorders, metastasis and malignancy, HIV (human immunodeficiency virus) infection and benign proliferative diseases. During ontogeny, SDF-1α is crucial for appropriate patterning, and it promotes growth and differentiation of critical cell types. During pancreatic regeneration, SDF-1α- CXCR4 (chemokine receptor) ligand-receptor pair is crucial for maintaining the duct cell survival, proliferation, and migration.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The stromal cell-derived factor-1alpha/CXCR4 ligand-receptor axis is critical for progenitor survival and migration in the pancreas.
Kayali AG et al
The Journal of Cell Biology, 163(4), 859-869 (2003)
Functional diversity of SDF-1 splicing variants.
Janowski M
Cell adhesion & migration, 3(3), 243-249 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service