Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6229

Sigma-Aldrich

PF-4/CXCL4 murine

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

CXCL4, Oncostatin A Ironplact, Platelet Factor-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

mouse

recombinant

expressed in E. coli

assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

10.3 kDa

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CXCL4(56744)

General description

PF-4 is a CXC chemokine that is expressed in megakaryocytes and stored in the α-granules of platelets. PF-4 is chemotactic towards neutrophils and monocytes and has been shown to inhibit angiogenesis. Recombinant murine PF-4 is an 8.2 kDa protein containing 76 amino acid residues, including the four highly conserved residues present in CXC chemokines.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service