Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6213

Sigma-Aldrich

sDLL-4 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Delta-like protein 4, Drosophila Delta homolog 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

54.3 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DLL-4(54567)

General description

Human sDLL4 comprises the extracellular signaling domain of DLL, a member of a structurally-related family of single-pass type I trans-membrane proteins that serve as ligands for Notch receptors.

Biochem/physiol Actions

DLL4 (delta like canonical Notch ligand 4) functions to specifically activate the Notch-1 and Notch-4 receptors. The Notch signaling pathway regulates endothelial-cell differentiation, proliferation and apoptosis, and is essential for the development, maintenance and remodeling of the vascular system. Targeted deletion of the DLL4 gene in mice resulted in severe vascular defects and death before birth. Up-regulation of DLL4 expression has been implicated in the vascular development of certain tumors. Recombinant human sDLL4 is a 54 160;kDa glycoprotein containing 498 amino-acid residues.[1]

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 1X PBS, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of endothelial cell proliferation by Notch1 signaling is mediated by repressing MAPK and PI3K/Akt pathways and requires MAML1.
Liu Z J, et al.
Faseb Journal, 20(7), 1009-1011 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service